Learn More
Bachem Trypanothione
SDP
Supplier:  Bachem 4014289.0005
Encompass_Preferred
Trypanothione, N1,N8-Bis(glutathionyl)-spermidine disulfide, 96304-42-6, C27H47N9O10S2, Mr 721.86, 5mg. Trypanothione (TS2) was purified from the insect trypanosomatid Crithidia fasciculata and identified as a covalently linked adduct of glutathione and spermidine. TS2 is utilized as a substrate by trypanothione reductase, a unique enzyme found in trypanosomes and leishmanias (Km = 51 uM).
Catalog No. 50-194-7478