Learn More
Quality Biological Inc Trypsin (0.05%) EDTA (0.1%), 100 mL
SDP
Catalog No. 507517415
Encompass_Preferred
Trypsin is a proteolytic enzyme ¿ Used to detach adherent cell from culture vessel surfaces. ¿ Typical use includes removing adherent cells from a culture surface. ¿ EDTA is often added to enhance enzymatic activity of trypsin solution. ¿ EDTA acts by neutralizing calcium and magnesium ions that enhance cell-to-cell adhesion. ¿ Trypsin, 0.05% ¿ EDTA, 0.1% assists in the dissociation of cell monolayers and tissue ¿ Most useful when dissociating fibroblasts and keratinocytes. ¿ Storage Conditions: -10¿C to -20¿C ¿ Shipping Condition: on dry ice ¿ Sterile Filtered ¿ Performance Tested ¿ Product and/or packaging contains recycled/renewable content ¿ Product and/or packaging is recyclable or reduces waste ¿ Product and/or packaging is safer to human and environmental health ¿ ¿ Max Shelf Life: 24 months
Catalog No. 50-751-7415 Supplier Quality Biological Inc Supplier No. 118087721EA
Trypsin is a proteolytic enzyme ¿ Used to detach adherent cell from culture vessel surfaces. ¿ Typical use includes removing adherent cells from a culture surface. ¿ EDTA is often added to enhance enzymatic activity of trypsin solution. ¿ EDTA acts by neutralizing calcium and magnesium ions that enhance cell-to-cell adhesion. ¿ Trypsin, 0.05% ¿ EDTA, 0.1% assists in the dissociation of cell monolayers and tissue ¿ Most useful when dissociating fibroblasts and keratinocytes. ¿ Storage Conditions: -10¿C to -20¿C ¿ Shipping Condition: on dry ice ¿ Sterile Filtered ¿ Performance Tested ¿ Product and/or packaging contains recycled/renewable content ¿ Product and/or packaging is recyclable or reduces waste ¿ Product and/or packaging is safer to human and environmental health ¿ ¿ Max Shelf Life: 24 months