Learn More
Electron Microscopy Sciences Trypsin 1:100, Powder 25 GR
SDP
Catalog No. 5029539
Encompass_Preferred
F.W. 372.09
Extract from Porcine Pancreas. Used for whole cell preparations. Activity >100 NF units/mg.
Catalog No. 50-295-39 Supplier Electron Microscopy Sciences Supplier No. 22200
F.W. 372.09
Extract from Porcine Pancreas. Used for whole cell preparations. Activity >100 NF units/mg.