Learn More
Bioworld Trypsin 1:250, 25 g
Catalog No. 50199001
Encompass_Preferred
Trypsin, 1:250 SpecificationsSource: Porcine pancreas Tissue Culture Grade Special preparation for single cell suspension Trypsin >250 N.F.U/mg Chymotrypsin <75 N.F. U/mg Moisture <5.0% Cell Disaggregation: passes
Catalog No. 50-199-001 Supplier Bioworld Supplier No. 215600321
Trypsin, 1:250 SpecificationsSource: Porcine pancreas Tissue Culture Grade Special preparation for single cell suspension Trypsin >250 N.F.U/mg Chymotrypsin <75 N.F. U/mg Moisture <5.0% Cell Disaggregation: passes