Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trypsin, 1-250 from Porcine Pancreas , MP Biomedicals, LLC

Catalog No. MP210313920 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
20 g
Packaging:
Glass
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
MP210313920 20 g Glass
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP210313920 Supplier MP Biomedicals, Inc Supplier No. 0210313920
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Trypsin, 1-250 from Porcine Pancreas has an activity >250 USP u/mg. Trypsin is a porcine pancreas-derived enzyme that is commonly used for the dissociation and disaggregation of anchorage-dependent mammalian cells and tissues.

  • The concentration of trypsin necessary to dislodge cells from their substrate is dependent primarily on the cell type and the age of the culture
  • The 1:250 designation indicates the tryptic activity based on standardized testing
  • This ratio indicates that one part trypsin will digest 250 parts casein in the USP activity assay
  • Trypsin is composed of two subunits, a-trypsin and b-trypsin. a-Trypsin is composed of two peptide chains and b-trypsin is composed of one chain
  • Trypsin is used to re-suspend cells adherent to the cell culture dish wall during the process of harvesting cells
  • Trypsin can also be used to dissociate dissected cells
  • Trypsin can be used to break down casein in breast milk
  • Trypsin is commonly used in biological research during proteomics experiments to digest proteins into peptides for mass spectrometry analysis
  • Trypsin can also be used to dissolve blood clots in its microbial form and treat inflammation in its pancreatic form
  • One USP trypsin unit is the activity causing a change in absorbance of 0.003 per minute under the conditions specified
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7-7-9002
Molecular Formula C35H47N7O10
Molecular Weight (g/mol) 725.80
InChI Key WGWZNYKOUXOZTC-UHFFFAOYNA-N
Synonym tryptase, trypsin, tryptase from human lung, tryptase from human lung, buffered aqueous solution, ≥5 units/mg protein, acetic acid; tert-butyl n-2s-1-2s-1-2s-5-diaminomethylideneamino-1-5-methyl-2-oxochromen-7-yl amino-1-oxopentan-2-yl amino-3-hydroxy-1-oxopropan-2-yl amino-1-oxo-3-phenylpropan-2-yl carbamate
PubChem CID 5311489

Specifications

Color White
pH ∼8.0
Packaging Glass
Quantity 20 g
Grade Not Available
Physical Form Powder
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.