Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Trypsin 1:250, porcine pancreas

Catalog No. AAJ6399318
Click to view available options
Quantity:
10 g
50 g

Catalog No. AAJ6399318

Supplier: Thermo Scientific Chemicals J6399318

Only null left
Add to Cart

Trypsin, CAS # 9002-07-7, is a digestive enzyme formed from trypsinogen in the pancreas. | CAS: 9002-07-7 | C35H47N7O10

Trypsin 1:250 is used in removing adherent cells from a culture surface. The concentration of trypsin necessary to dislodge cells from their substrate is dependent primarily on the cell type and the age of the culture. It has also been used for the re-suspension of cells during cell culture, in proteomics research for digestion of proteins and in various in-gel digestions. Additional applications include assessing crystallization by membrane-based techniques and in a study to determine that protein folding rates and yields can be limited by the presence of kinetic traps.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Trypsin is found in the digestive system of vertebrates
  • Trypsin is a serine protease that hydrolyzes the carboxyl end of amino acids lysine and arginine
  • Trypsin is produced in the pancreas from trypsinogen and is released to downstream polypeptides in the small intestine

Application

  • Trypsin can be used to resuspend adherent cells for harvesting in cell culture applications
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7-7-9002
Molecular Formula C35H47N7O10
Synonym EC 3.4.21.4

Specifications

For Use With (Application) A pancreatic serine protease
Content And Storage -30°C to -10°C
Form Powder
Quantity 50 g
Product Type Trypsin

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.