Learn More
FUJIFILM Biosciences Trypsin 1250 2.5 10X sol
Catalog No. 5000450262
Encompass
Trypsin Solution 10X is an enzyme preparation used to remove cells from the culture vessel surface into suspension This formulation is a 2 5% solution of 1250 trypsin in normal saline
Catalog No. 50-004-50262 Supplier FUJIFILM Biosciences Supplier No. 9336100ML
Trypsin Solution 10X is an enzyme preparation used to remove cells from the culture vessel surface into suspension This formulation is a 2 5% solution of 1250 trypsin in normal saline