Learn More
Worthington Biochemical Corporation TRYPSIN 2X 100MG
SDP
Catalog No. NC9185815
Encompass
Trypsin, Code: TRL, Minimum activity: 180U/mg Protein, Supplied as a chromatographically purified, diafiltered and lyophilized powder, TPCK treated. Store at 2 deg. to 8 deg.C, Protect from moisture, 100mg
Catalog No. NC9185815 Supplier Worthington Biochemical Corporation Supplier No. LS003702
Trypsin, Code: TRL, Minimum activity: 180U/mg Protein, Supplied as a chromatographically purified, diafiltered and lyophilized powder, TPCK treated. Store at 2 deg. to 8 deg.C, Protect from moisture, 100mg