Learn More
Sigma Aldrich Fine Chemicals Biosciences Trypsin Acetylated bo100MG
Catalog No. 502900923
Encompass
The trypsin molecule has two domains one is related to the enzyme active site and the tryptophan residues the other is related to the 8-anilinonaphthalene-1-sulfonate binding.
Catalog No. 50-290-0923 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. T6763100MG
The trypsin molecule has two domains one is related to the enzyme active site and the tryptophan residues the other is related to the 8-anilinonaphthalene-1-sulfonate binding.