Learn More
Sigma Aldrich Fine Chemicals Biosciences Trypsin from bovine pancreas TPCK Treated, essentially salt-free, lyophilized powder, >=10,000 BAEE units/mg protein | 9002-07-7 | MFCD00082094 | 250MG
Catalog No. NC0664002
Encompass
Trypsin from bovine pancreas TPCK Treated, essentially salt-free, lyophilized powder, >=10,000 BAEE units/mg protein | 9002-07-7 | MFCD00082094 | 250MG
Catalog No. NC0664002 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. T1426250MG
Trypsin from bovine pancreas TPCK Treated, essentially salt-free, lyophilized powder, >=10,000 BAEE units/mg protein | 9002-07-7 | MFCD00082094 | 250MG