Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Quality Biological Inc Trypsin Delarco, pH 6.8, Pack 4 X 100 mL
SDP

Catalog No. 509840119
Encompass_Preferred

Trypsin Delarco assists in the process of cell culture and transfection. Trypsin is a proteolytic enzyme used to detach adherent cells from culture vessel surfaces. Trypsin delarco is used in cell passaging, to ensure stable products.

With HBSS
Without Calcium and Magnesium
Without Phenol Red

Storage Conditions: -10 °C to -20 °C
Shipping Condition: on dry ice
Sterile Filtered
pH: 6.8
Concentration: 0.1 mM disodium EDTA
Max Shelf Life: 36 months from date of manufacture

Manufactured under an ISO 9001:2015 certified quality management system.

Product and/or packaging contains recycled/renewable content. Product and/or packaging is recyclable or reduces waste. Product and/or packaging is safer to human and environmental health.

Catalog No. 50-984-0119 Supplier Quality Biological Inc Supplier No. 118150721
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Trypsin Delarco assists in the process of cell culture and transfection. Trypsin is a proteolytic enzyme used to detach adherent cells from culture vessel surfaces. Trypsin delarco is used in cell passaging, to ensure stable products.

With HBSS
Without Calcium and Magnesium
Without Phenol Red

Storage Conditions: -10 °C to -20 °C
Shipping Condition: on dry ice
Sterile Filtered
pH: 6.8
Concentration: 0.1 mM disodium EDTA
Max Shelf Life: 36 months from date of manufacture

Manufactured under an ISO 9001:2015 certified quality management system.

Product and/or packaging contains recycled/renewable content. Product and/or packaging is recyclable or reduces waste. Product and/or packaging is safer to human and environmental health.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.