Learn More
STEMCELL Technologies Trypsin-EDTA (0.05%), Size: 500 mL
SDP
Catalog No. NC2031805
Encompass
Trypsin-EDTA (0.05%) can be used to achieve a single-cell suspension of human embryonic stem (ES) cells or human induced pluripotent stem (iPS) cells for flow cytometry or cloning assays. This is a gentler version than the standard concentration (0.25%), and has applications to many other cell culture systems in addition to human ES cells and iPS cells.
Catalog No. NC2031805 Supplier STEMCELL Technologies Supplier No. 07910
Trypsin-EDTA (0.05%) can be used to achieve a single-cell suspension of human embryonic stem (ES) cells or human induced pluripotent stem (iPS) cells for flow cytometry or cloning assays. This is a gentler version than the standard concentration (0.25%), and has applications to many other cell culture systems in addition to human ES cells and iPS cells.