Learn More
STEMCELL Technologies Trypsin-EDTA (0.25%), Size: 500 mL
SDP
Catalog No. NC2031804
Encompass
Trypsin-EDTA (0.25%) is recommended for the dissociation of adherent cells, cell aggregates, and tissues into single-cell suspensions.
Catalog No. NC2031804 Supplier STEMCELL Technologies Supplier No. 07901
Trypsin-EDTA (0.25%) is recommended for the dissociation of adherent cells, cell aggregates, and tissues into single-cell suspensions.