Learn More
GenDEPOT TRYPSIN-EDTA(10X), 0.5%, 100ML
Catalog No. 501015446
Encompass_Preferred
Trypsin solutions are used widely for dissociation of tissues and cell monolayers and we can deliver solutions that will perform consistently, batch after batch. The biochemical assays performed on Trypsin 1:250 determine both trypsin specific-activity at the level of certain co-purified enzymes that influence cell removal and viability. All trypsin solutions are prepared from porcine-parvovirus-tested and mycoplasma-tested materials. Formulation Contains 0.5% trypsin, 5.3mM EDTA , and 8.5 g/L NaCl. No phenol red. Sterile filtered.
Catalog No. 50-101-5446 Supplier GenDEPOT Supplier No. CA015010
Trypsin solutions are used widely for dissociation of tissues and cell monolayers and we can deliver solutions that will perform consistently, batch after batch. The biochemical assays performed on Trypsin 1:250 determine both trypsin specific-activity at the level of certain co-purified enzymes that influence cell removal and viability. All trypsin solutions are prepared from porcine-parvovirus-tested and mycoplasma-tested materials. Formulation Contains 0.5% trypsin, 5.3mM EDTA , and 8.5 g/L NaCl. No phenol red. Sterile filtered.