Learn More
Promocell GmbH PromoCell Trypsin / EDTA 125 ml, 0.04 % Trypsin / 0.03 % EDTA
Catalog No. 50306375
Encompass
Trypsin is still the most widely employed enzyme used for cell detachment. PromoCell´s Trypsin Solutions exhibit standardized enzyme activity and contain trypsin from extra-pure lots.Trypsin / EDTA Solution having a ratio of 0.04% / 0.03%.
Catalog No. 50-306-375 Supplier Promocell GmbH Supplier No. C41010
Trypsin is still the most widely employed enzyme used for cell detachment. PromoCell´s Trypsin Solutions exhibit standardized enzyme activity and contain trypsin from extra-pure lots.Trypsin / EDTA Solution having a ratio of 0.04% / 0.03%.