Learn More
Promocell GmbH PromoCell Trypsin / EDTA 2 250 ml, 0,025 % Trypsin / 0,01 % EDTA
Supplier:  Promocell GmbH C41022
Encompass
Trypsin is still the most widely employed enzyme used for cell detachment. PromoCell´s Trypsin Solutions exhibit standardized enzyme activity and contain trypsin from extra-pure lots.Trypsin / EDTA Solution having a ratio of 0.025% / 0.01%.
Catalog No. 50-306-378