Learn More
Lifeline Cell Technology Trypsin 0.05% EDTA 0.02% (phenol red free), 100mL
SDP
Catalog No. NC9972660
Encompass_Preferred
Trypsin acts as an enzyme to help dissociate cells from their culture vessel. EDTA is a calcium chelator that helps to prevent clumping of cells. 0.05% Trypsin/0.02% EDTA Solution may be stored at -20°C until the expiration date or at 4-8°C for up to 1 month. This product is not approved for human or veterinary use or for use in in vitro diagnostics or clinical procedures.
Catalog No. NC9972660 Supplier Lifeline Cell Technology Supplier No. CM0017
Trypsin acts as an enzyme to help dissociate cells from their culture vessel. EDTA is a calcium chelator that helps to prevent clumping of cells. 0.05% Trypsin/0.02% EDTA Solution may be stored at -20°C until the expiration date or at 4-8°C for up to 1 month. This product is not approved for human or veterinary use or for use in in vitro diagnostics or clinical procedures.