Learn More
Lonza Walkersville Trypsin/EDTA for Mesenchymal Stem Cells
Catalog No. NC9988096
Encompass
Trypsin/EDTA (catalog no. CC-3232): is Lonza's recommended Trypsin/EDTA for all strongly adherent primary cells including normal human articular chondrocytes (NHAC-Kn), human bone marrow derived mesenchymal stem cells (hMSC), and dental pulp stem cells (DPSC).
Catalog No. NC9988096 Supplier Lonza Walkersville Supplier No. CC3232
Trypsin/EDTA (catalog no. CC-3232): is Lonza's recommended Trypsin/EDTA for all strongly adherent primary cells including normal human articular chondrocytes (NHAC-Kn), human bone marrow derived mesenchymal stem cells (hMSC), and dental pulp stem cells (DPSC).