Learn More
Lonza Walkersville Trypsin/EDTA Solution, 100 mL
Catalog No. NC9533708
Encompass
Trypsin is a clear, colorless-yellow, highly active bovine enzyme preparation, used to remove cells from the culture vessel surface into suspension.It is generally used at a working concentration of 0.025%, prepared in a balanced salt solution, without calcium or magnesium at a pH of 7.6-7.8.After treatment, trypsin activity can be halted by adding a serum media mixture or a trypsin inhibitor (from soybean, for example) in serum-free systems.Trypsin/EDTA is also a component of Lonza's ReagentPack Subculture Reagents kit (CC-5034) which includes Trypsin Neutralizing reagent and HEPES buffer.Content:1 x Bottle Trypsin/EDTA 100 mL
Catalog No. NC9533708 Supplier Lonza Walkersville Supplier No. CC5012
Trypsin is a clear, colorless-yellow, highly active bovine enzyme preparation, used to remove cells from the culture vessel surface into suspension.It is generally used at a working concentration of 0.025%, prepared in a balanced salt solution, without calcium or magnesium at a pH of 7.6-7.8.After treatment, trypsin activity can be halted by adding a serum media mixture or a trypsin inhibitor (from soybean, for example) in serum-free systems.Trypsin/EDTA is also a component of Lonza's ReagentPack Subculture Reagents kit (CC-5034) which includes Trypsin Neutralizing reagent and HEPES buffer.Content:1 x Bottle Trypsin/EDTA 100 mL