Learn More
Sigma Aldrich Fine Chemicals Biosciences Trypsin-EDTA solution 0.25%, sterile-filtered, BioReagent, suitable for cell culture, 2.5 g porcine trypsin and 0.2 g EDTA . 4Na per liter of Hanks' Balanced Salt Solution with phenol red | MFCD00130286 | 500ML
Catalog No. NC2061733
Encompass
Trypsin-EDTA solution 0.25%, sterile-filtered, BioReagent, suitable for cell culture, 2.5 g porcine trypsin and 0.2 g EDTA . 4Na per liter of Hanks' Balanced Salt Solution with phenol red | MFCD00130286 | 500ML
Catalog No. NC2061733 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. T4049500ML
Trypsin-EDTA solution 0.25%, sterile-filtered, BioReagent, suitable for cell culture, 2.5 g porcine trypsin and 0.2 g EDTA . 4Na per liter of Hanks' Balanced Salt Solution with phenol red | MFCD00130286 | 500ML