Learn More
Sigma Aldrich Fine Chemicals Biosciences Trypsin-EDTA solution 1 x, sterile; sterile-filtered, BioReagent, suitable for cell culture, 0.5 g porcine trypsin and 0.2 g EDTA . 4Na per liter of Hanks' Balanced Salt Solution with phenol red | MFCD00130286 | 100ML
Supplier:  Sigma Aldrich Fine Chemicals Biosciences T3924100ML
Encompass
Trypsin-EDTA solution 1 x, sterile; sterile-filtered, BioReagent, suitable for cell culture, 0.5 g porcine trypsin and 0.2 g EDTA . 4Na per liter of Hanks' Balanced Salt Solution with phenol red | MFCD00130286 | 100ML
Catalog No. NC0043665