Learn More
Sigma Aldrich Fine Chemicals Biosciences Trypsin-EDTA solution, MFCD00130286, 500 mL
Supplier:  Sigma Aldrich Fine Chemicals Biosciences T3924500ML
Encompass
1 ×, sterile; sterile-filtered, BioReagent, suitable for cell culture, 0.5 g porcine trypsin and 0.2 g EDTA • 4Na per liter of Hanks′ Balanced Salt Solution with phenol red. The typical use for this product is in removing adherent cells from a culture surface. The concentration of trypsin necessary to dislodge cells from their substrate is dependent primarily on the cell type and the age of the culture.
Catalog No. NC0067125