Learn More
Abcam Trypsin Antigen Retrieval Solution, 25ML
Catalog No. NC0653890
Encompass_Preferred
Trypsin-based antigen retrieval solution for Protease-Induced Epitope Retrieval (PIER) (ab970).
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC0653890 Supplier Abcam Supplier No. AB97025ML
Trypsin-based antigen retrieval solution for Protease-Induced Epitope Retrieval (PIER) (ab970).
The product is subject to the following: Abcam Restricted Use Statement