Learn More
Medchemexpress LLC Trypsin, Ethylenediaminetetraacetic acid | 9002-07-7 | MFCD00082094 | 100 ML
SDP
Catalog No. 5000429808
Encompass_Preferred
MCE 0.25% Trypsin-EDTA (1x), Phenol Red-Free, is a solution prepared by dissolving trypsin powder (a mixture of proteases from porcine pancreas, irradiated and sterilized) and EDTA in a calcium- and magnesium-free balanced salt solution. It is filter-sterilized and can be directly used for the dissociation of cultured cells and tissues. The 100 mL is defined as the base specification. All larger sizes correspond to incremental volumes of this base.
  • Trypsin hydrolyzes intercellular proteins and disperses primary tissues or adherent cells into individual cells.
  • Can be widely used for cell dissociation, cell culture passaging and primary tissue dissociation.
  • Prepared with trypsin powder and EDTA in a calcium- and magnesium-free balanced salt solution.
  • Filter-sterilized.
  • Can be directly used for the dissociation of cultured cells and tissues.
  • Phenol Red-Free.
Catalog No. 50-004-29808 Supplier Medchemexpress LLC Supplier No. HYK3009100ML
MCE 0.25% Trypsin-EDTA (1x), Phenol Red-Free, is a solution prepared by dissolving trypsin powder (a mixture of proteases from porcine pancreas, irradiated and sterilized) and EDTA in a calcium- and magnesium-free balanced salt solution. It is filter-sterilized and can be directly used for the dissociation of cultured cells and tissues. The 100 mL is defined as the base specification. All larger sizes correspond to incremental volumes of this base.
  • Trypsin hydrolyzes intercellular proteins and disperses primary tissues or adherent cells into individual cells.
  • Can be widely used for cell dissociation, cell culture passaging and primary tissue dissociation.
  • Prepared with trypsin powder and EDTA in a calcium- and magnesium-free balanced salt solution.
  • Filter-sterilized.
  • Can be directly used for the dissociation of cultured cells and tissues.
  • Phenol Red-Free.