Learn More
STEMCELL Technologies Trypsin in Citrate Saline 0.25, 100 mL
SDP
Catalog No. 502572474
Encompass
0.25% porcine trypsin in citrate saline solution for the detachment of adherent cells
Catalog No. 50-257-2474 Supplier STEMCELL Technologies Supplier No. 07400
0.25% porcine trypsin in citrate saline solution for the detachment of adherent cells