Learn More
Worthington Biochemical Corporation TRYPSIN INHBITOR OVOMU 2GM
SDP
Supplier:  Worthington Biochemical Corporation LS003086
Encompass
Mucoprotein and antitryptic factor of egg white described by Lineweaver and Murray, J. Biol. Chem., 171, 565 (1947). A dialyzed, dried powder. 9035-81-8
Catalog No. 50-592-358