Learn More
Sigma Aldrich Fine Chemicals Biosciences Trypsin Inhibitor from soy
Catalog No. 502901865
Encompass
Soybean trypsin inhibitor (STI) comprises 181 amino acids. It is a monomeric protein corresponding to a molecular weight of 20 kDa and has two disulfide bridges. It belongs to serpins A serine protease inhibitors family.
Catalog No. 50-290-1865 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 10109886001
Soybean trypsin inhibitor (STI) comprises 181 amino acids. It is a monomeric protein corresponding to a molecular weight of 20 kDa and has two disulfide bridges. It belongs to serpins A serine protease inhibitors family.