Learn More
Crescent Chemical Co Inc TRYPSIN INHIBITOR FROM SOYBEAN
SDP
Catalog No. 50248259
Encompass_Preferred
Trypsin inhibitor from soybean min. 13 000 U/mg lyophil. Salt-free. Trypsin is inhibited in a molar ratio of 1:1. Inhibits as well chymotrypsin (2), plasmin, kallikrein, thrombin and other proteolytic enzymes. Size - 250MG Storage Conditions - +2 °C TO +8 °C Catalog Number - 37329
Catalog No. 50-248-259 Supplier Crescent Chemical Co Inc Supplier No. SE37329.01
Trypsin inhibitor from soybean min. 13 000 U/mg lyophil. Salt-free. Trypsin is inhibited in a molar ratio of 1:1. Inhibits as well chymotrypsin (2), plasmin, kallikrein, thrombin and other proteolytic enzymes. Size - 250MG Storage Conditions - +2 °C TO +8 °C Catalog Number - 37329