Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trypsin inhibitor, MP Biomedicals™

Catalog No. ICN10111390 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
25 mg
100 mg
250 mg
500 mg
1 g
5 g
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10111390 500 mg
ICN10111325 25 mg
ICN10111380 100 mg
ICN10111383 250 mg
ICN10111391 1 g
ICN10111394 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. ICN10111390 Supplier MP Biomedicals, Inc Supplier No. 0210111390
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 9035-81-8 | Weight: 21500 g/mol

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9035-81-8
Synonym Ovomucoid trypsin inhibitor

Specifications

Color White
Quantity 500 mg
Physical Form Powder
Chemical Name or Material Trypsin Inhibitor
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.