Learn More
Worthington Biochemical Corporation Trypsin Inhibitor, Soybean, Purified 1gm
SDP
Supplier:  Worthington Biochemical Corporation LS003571
Encompass
SoybeanAnimal Free/AF. Chromatographically purified as described by Frattali, V., and Steiner, R.: Biochem., 7, 521 (1968), modified by Worthington. A dialyzed, lyophilized powder. Purity >90% checked using SDS PAGE.Store at 2-8°C.
Catalog No. NC0104269