Learn More
Cayman Chemical TrypsIn InhIbItrsoyben 100mg
SDP
Catalog No. 502968820
Encompass_Preferred
An inhibitor of serine proteases that dose-dependently inhibits trypsin from a variety of sources, as well as plasma kallikrein, thrombin, plasmin, and other serine proteases; partially inhibits chymotrypsin and neutrophil elastase
Catalog No. 50-296-8820 Supplier Cayman Chemical Supplier No. 14502100 MG
An inhibitor of serine proteases that dose-dependently inhibits trypsin from a variety of sources, as well as plasma kallikrein, thrombin, plasmin, and other serine proteases; partially inhibits chymotrypsin and neutrophil elastase