Learn More
Cayman Chemical TrypsIn InhIbItrsoyben 250mg
SDP
Supplier:  Cayman Chemical 14502250 MG
Encompass_Preferred
An inhibitor of serine proteases that dose-dependently inhibits trypsin from a variety of sources, as well as plasma kallikrein, thrombin, plasmin, and other serine proteases; partially inhibits chymotrypsin and neutrophil elastase
Catalog No. 50-297-1802