Learn More
Electron Microscopy Sciences Trypsin Kit Trypsin Kit Large
SDP
Catalog No. 501926520
Encompass_Preferred
Trypsin is used for proteolytic digestion of formalin-fixed paraffin-embedded (FFPE) tissue sections prior to application of antibodies. In IHC most commonly used fixative like formalin; mask tissue antigens (cellular, membrane and nuclear) by cross-linking process, this results in poor or no staining in IHC. Trypsin digestion improves immunoreactivity of some antigens in FFPE tissue sections. For cytokeratin clone AE3 and AE1/AE3, this enzyme works much better than boiling the tissue with citrate buffer.
Intended Use: Use as antigen retriever for some antibodies in IHC. Please refer to primary antibody protocol for IHC.
Reagent: This enzyme is supplied as concentrated form along with buffer. Reagent B (Buffer) is 100ml and Reagent E (Enzyme) is 10ml.
Catalog No. 50-192-6520 Supplier Electron Microscopy Sciences Supplier No. 6414202
Trypsin is used for proteolytic digestion of formalin-fixed paraffin-embedded (FFPE) tissue sections prior to application of antibodies. In IHC most commonly used fixative like formalin; mask tissue antigens (cellular, membrane and nuclear) by cross-linking process, this results in poor or no staining in IHC. Trypsin digestion improves immunoreactivity of some antigens in FFPE tissue sections. For cytokeratin clone AE3 and AE1/AE3, this enzyme works much better than boiling the tissue with citrate buffer.
Intended Use: Use as antigen retriever for some antibodies in IHC. Please refer to primary antibody protocol for IHC.
Reagent: This enzyme is supplied as concentrated form along with buffer. Reagent B (Buffer) is 100ml and Reagent E (Enzyme) is 10ml.