Learn More
Crescent Chemical Co Inc TRYPSIN NB SEQUENCING GRADE
SDP
Catalog No. 50248247
Encompass_Preferred
Trypsin NB sequencing grade, modified from porcine pancreas Modified by reductive methylation, no chymotryptic activity detectable.Specificity verified by digestion of oxidized B-chain of insulin.Trypsin NB Sequencing Grade, modified, is suitable for digestion of proteins for mass spectrometry analysis. R
Catalog No. 50-248-247 Supplier Crescent Chemical Co Inc Supplier No. SE37283.01
Trypsin NB sequencing grade, modified from porcine pancreas Modified by reductive methylation, no chymotryptic activity detectable.Specificity verified by digestion of oxidized B-chain of insulin.Trypsin NB Sequencing Grade, modified, is suitable for digestion of proteins for mass spectrometry analysis. R