Learn More
Lonza Walkersville Trypsin Neutralizing Solution, 100 mL
Catalog No. NC9533709
Encompass
Trypsin Neutralizing Solution (TNS) is used to effectively neutralize Trypsin/EDTA. Lonza's TNS is availableindividually or as a component in our ReagentPack Subculture Reagents (CC-5034) which includes Trypsin and HEPES buffer.Content:1 x Bottle Trypsin Neutralizing Solution, 100 mL
Catalog No. NC9533709 Supplier Lonza Walkersville Supplier No. CC5002
Trypsin Neutralizing Solution (TNS) is used to effectively neutralize Trypsin/EDTA. Lonza's TNS is availableindividually or as a component in our ReagentPack Subculture Reagents (CC-5034) which includes Trypsin and HEPES buffer.Content:1 x Bottle Trypsin Neutralizing Solution, 100 mL