Learn More
ATCC Trypsin Neutralizing Solution
Supplier:  ATCC PCS999004
Encompass
Trypsin Neutralizing Solution is specifically formulated (5% FBS in phosphate-buffered saline without calcium and magnesium) to rapidly inactivate the concentration of trypsin found in the Trypsin-EDTA for Primary Cells solution.
Catalog No. 50-238-4117