Learn More
ILEX LIFE SCIENCES LLC TRYPSIN P10-021500 500ML
SDP
Catalog No. 502733683
Encompass_Preferred
PAN-Biotech Trypsin 0.25% in PBS, w/o: Ca and Mg (500 ml). DESCRIPTION: PAN-Biotech Trypsin 0.25% in PBS, w/o: Ca and Mg is a porcine pancreas-derived enzyme in PBS solution. It is commonly used for the dissociation of anchorage-dependent (adherent) mammalian cells and tissues from culture surfaces. The concentration of trypsin necessary to dislodge cells from their substrate is dependent on the cell type and the age of the culture. APPLICATION: Enzymatic solution for the dissociation and disaggregation of anchorage-dependent mammalian cells and tissues. Trypsin solutions can range from 0.025% to 0.5% depending on application. LIQUID/POWDER: Liquid. PHENOL RED: Without Phenol Red. Ca/Mg CONTENT: Without Ca and Mg. TRYPSIN: 0.25% Trypsin. EDTA: No EDTA. STERILE: Yes. STORAGE TEMP: -20°C. MANUFACTURER: PAN-Biotech GmbH (Germany).
Catalog No. 50-273-3683 Supplier ILEX LIFE SCIENCES LLC Supplier No. P10021500
Add to Cart
Add to Cart
PAN-Biotech Trypsin 0.25% in PBS, w/o: Ca and Mg (500 ml). DESCRIPTION: PAN-Biotech Trypsin 0.25% in PBS, w/o: Ca and Mg is a porcine pancreas-derived enzyme in PBS solution. It is commonly used for the dissociation of anchorage-dependent (adherent) mammalian cells and tissues from culture surfaces. The concentration of trypsin necessary to dislodge cells from their substrate is dependent on the cell type and the age of the culture. APPLICATION: Enzymatic solution for the dissociation and disaggregation of anchorage-dependent mammalian cells and tissues. Trypsin solutions can range from 0.025% to 0.5% depending on application. LIQUID/POWDER: Liquid. PHENOL RED: Without Phenol Red. Ca/Mg CONTENT: Without Ca and Mg. TRYPSIN: 0.25% Trypsin. EDTA: No EDTA. STERILE: Yes. STORAGE TEMP: -20°C. MANUFACTURER: PAN-Biotech GmbH (Germany).