Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Trypsin, porcine pancreas

Catalog No. AAJ6040209
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6040209 10 g
AAJ6040218 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6040209 Supplier Thermo Scientific Chemicals Supplier No. J6040209
Add to Cart

missing translation for 'validMainframeMsg'

A pancreatic serine protease | CAS: 9002-07-7 | C35H47N7O10 | 23.8 kd g/mol

Trypsin, suitable for use in ELISA or for antigen applications in immunological protocols. Other applications include it is been used for the research purpose.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Reconstitute in aqueous buffer
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7-7-9002
Molecular Formula C35H47N7O10
Synonym EC 3.4.21.4; Trypsin IV porcine

Specifications

For Use With (Application) A pancreatic serine protease
Molecular Weight (g/mol) 23.8 kd
Content And Storage -30°C to -10°C
Form Lyophilized Powder
Quantity 10 g
Product Type Trypsin

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.