Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Trypsin, Porcine Pancreas

Catalog No. ICN15357180 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15357180 100 g
ICN15357125 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN15357180 Supplier MP Biomedicals™ Supplier No. 0215357180
Add to Cart
Add to Cart

Typical use for this product is in removing adherent cells from a culture surface. The concentration of trypsin necessary to dislodge cells from their substrate is dependent primarily on the cell type and the age of the culture.

  • CAS Number: 9002-07-7
  • Molecular Weight: Approximately 23,000 or 22000 (for human)
  • Composition: Trypsin is composed of two subunits, a-trypsin and b-trypsin. a-Trypsin is composed of two peptide chains and b-trypsin is composed of one chain.
  • Activity: One BAEE unit will produce a DA253 of 0.001 per minute at pH 7.6 at 25°C using BAEE as substrate. Reaction volume = 3.2mL (1 cm light path).
    One TAME unit hydrolyzes 1 umole of p-toluene-sulfonyl-L-arginine methyl ester (TAME) per minute at 25°C, pH 8.2, in the presence of 0.001 M calcium ion.
    One USP trypsin unit is the activity causing a change in absorbance of 0.003 per minute under the conditions specified.
  • Activity Conversion:1 TAME unit = 19.2 USP or NF units = 57.5 BAEE Units
  • Composition: Trypsin is composed of two subunits, a-trypsin and b-trypsin. a-Trypsin is composed of two peptide chains and b-trypsin is composed of one chain.
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Release adherent cells from tissue culture plates for passaging
Sample Type Porcine
Content And Storage 4°C
Form Powder
Quantity 100 g
Product Type Trypsin
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.