Learn More
Sigma Aldrich Fine Chemicals Biosciences Trypsin recombinant Proteo
Catalog No. 502901552
Encompass
This trypsin preparation is generated from the recombinant Pichia pastoris strain and is thus free of chymotrypsin. Trypsin recombinant Proteomics Grade exhibits high specific activity.
Catalog No. 50-290-1552 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 3708985001
This trypsin preparation is generated from the recombinant Pichia pastoris strain and is thus free of chymotrypsin. Trypsin recombinant Proteomics Grade exhibits high specific activity.