Learn More
Sigma Aldrich Fine Chemicals Biosciences Trypsin Sequencing Grade m
Catalog No. 502891536
Encompass
Trypsin Sequencing Grade, modified, is isolated from bovine pancreas as a highly purified and specific protease, and subsequently modified.Inhibitors TLCK, DFP, PMSF, leupeptin, soybean trypsin inhibitor, trypsin inhibitor from hen egg, aprotinin, alpha2-macroglobulin,alpha1-antitrypsin, APMSF, and antipain.
Catalog No. 50-289-1536 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 11418025001
Trypsin Sequencing Grade, modified, is isolated from bovine pancreas as a highly purified and specific protease, and subsequently modified.Inhibitors TLCK, DFP, PMSF, leupeptin, soybean trypsin inhibitor, trypsin inhibitor from hen egg, aprotinin, alpha2-macroglobulin,alpha1-antitrypsin, APMSF, and antipain.