Learn More
Worthington Biochemical Corporation TRYPSIN TPCK TREATED 1G
SDP
Catalog No. NC9211549
Encompass
A chromatographically purified, diafiltered, lyophilized powder that has been treated with L-(tosylamido-2-phenyl) ethyl chloromethyl ketone (TPCK) to inhibit contaminating chymotryptic activity [Kostka and Carpenter, J. Biol. Chem. 239, 1799 (1964)].
Catalog No. NC9211549 Supplier Worthington Biochemical Corporation Supplier No. LS003744
A chromatographically purified, diafiltered, lyophilized powder that has been treated with L-(tosylamido-2-phenyl) ethyl chloromethyl ketone (TPCK) to inhibit contaminating chymotryptic activity [Kostka and Carpenter, J. Biol. Chem. 239, 1799 (1964)].