Learn More
Worthington Biochemical Corporation Trypsin Vial, NCIS, 5 vials
SDP
Catalog No. NC9300430
Encompass
A component of the NCIS kit. This material is 0.22 micron membrane filtered and lyophilized in autoclaved vials. A vial reconstituted with 2 ml of HBSS yields a solution of 500µg/ml of trypsin, Code: TRLS. Suitable for cell isolation and culture applications. Store at 2-8°C.
Catalog No. NC9300430 Supplier Worthington Biochemical Corporation Supplier No. LK003225
A component of the NCIS kit. This material is 0.22 micron membrane filtered and lyophilized in autoclaved vials. A vial reconstituted with 2 ml of HBSS yields a solution of 500µg/ml of trypsin, Code: TRLS. Suitable for cell isolation and culture applications. Store at 2-8°C.