Learn More
Bioworld Trypsin (Bovine), 1 g
Supplier:  Bioworld 215600311
Encompass_Preferred
Bovine Trypsin A serine peptidase that cleaves at internal peptide bonds on the carboxyl-side of arginine and lysine residues. >USP Grade Activity (min.) at 25 C >2,500U/mg Chymotrypsin 5.0% Unit Definition: The amount of enzyme which causes an increase in absorbance of 0.003 per minute at 25 C and 253 nm, pH 7.6 on the substrate benzoyl-L-arginine ethyl ester (BTEE). 1 gram
Catalog No. 50-198-999