Learn More
ATCC Trypsin/EDTA, 1X, 100 ML
Catalog No. 502384982
Encompass
The Trypsin-EDTA Solution is 0.25% Trypsin/0.53 mM EDTA in Hanks Balanced Salt Solution without calcium or magnesium. For dissociation of cell monolayers. Trypsin-EDTA solution is suitable for most but not for all adherent cell lines. This product has applications for cell culture, cell growth, and viability. For cell line-specific information, please go to the appropriate product page on the web, refer to the product sheet supplied with the cell line, or contact ATCC Technical Service.
Catalog No. 50-238-4982 Supplier ATCC Supplier No. 302101
The Trypsin-EDTA Solution is 0.25% Trypsin/0.53 mM EDTA in Hanks Balanced Salt Solution without calcium or magnesium. For dissociation of cell monolayers. Trypsin-EDTA solution is suitable for most but not for all adherent cell lines. This product has applications for cell culture, cell growth, and viability. For cell line-specific information, please go to the appropriate product page on the web, refer to the product sheet supplied with the cell line, or contact ATCC Technical Service.