Learn More
ILEX LIFE SCIENCES LLC TRYPSINEDTA P10-020500 6X500ML
SDP
Catalog No. 502733686
Encompass_Preferred
PAN-Biotech Trypsin 0.25% / EDTA 0.02% in DPBS, w/o: Ca and Mg (6 x 500 ml). DESCRIPTION: PAN-Biotech Trypsin 0.25% / EDTA 0.02% in PBS, w/o: Ca and Mg is a porcine pancreas-derived enzyme plus EDTA in a PBS solution. Trypsin-EDTA solution is commonly used for the dissociation of anchorage-dependent (adherent) mammalian cells and tissues from culture surfaces. The concentration of trypsin necessary to dislodge cells from their substrate is dependent on the cell type and the age of the culture. Ethylenediaminetetraacetic acid (EDTA), a chelating agent, is often added to trypsin solutions to enhance enzymatic activity by neutralizing calcium and magnesium ions that enhance cell-to-cell adhesion and obscure the peptide bonds on which trypsin acts. LIQUID/POWDER: Liquid. PHENOL RED: Without Phenol Red. Ca/Mg CONTENT: Without Ca and Mg. TRYPSIN: 0.25% Trypsin. EDTA: 0.02% EDTA. STERILE: Yes. STORAGE TEMP.: -20°C. MANUFACTURER: PAN-Biotech GmbH (Germany).
Catalog No. 50-273-3686 Supplier ILEX LIFE SCIENCES LLC Supplier No. P10020500X6
Add to Cart
Add to Cart
PAN-Biotech Trypsin 0.25% / EDTA 0.02% in DPBS, w/o: Ca and Mg (6 x 500 ml). DESCRIPTION: PAN-Biotech Trypsin 0.25% / EDTA 0.02% in PBS, w/o: Ca and Mg is a porcine pancreas-derived enzyme plus EDTA in a PBS solution. Trypsin-EDTA solution is commonly used for the dissociation of anchorage-dependent (adherent) mammalian cells and tissues from culture surfaces. The concentration of trypsin necessary to dislodge cells from their substrate is dependent on the cell type and the age of the culture. Ethylenediaminetetraacetic acid (EDTA), a chelating agent, is often added to trypsin solutions to enhance enzymatic activity by neutralizing calcium and magnesium ions that enhance cell-to-cell adhesion and obscure the peptide bonds on which trypsin acts. LIQUID/POWDER: Liquid. PHENOL RED: Without Phenol Red. Ca/Mg CONTENT: Without Ca and Mg. TRYPSIN: 0.25% Trypsin. EDTA: 0.02% EDTA. STERILE: Yes. STORAGE TEMP.: -20°C. MANUFACTURER: PAN-Biotech GmbH (Germany).