Learn More
ILEX LIFE SCIENCES LLC TRYPSINEDTA P10-0231SP 6X100ML
SDP
Supplier:  ILEX LIFE SCIENCES LLC P100231SPX6
Encompass_Preferred
PAN-Biotech Trypsin 0.05% / EDTA 0.02% in DPBS, w/o: Ca and Mg, w: Phenol red (6 x 100 ml). DESCRIPTION: PAN-Biotech Trypsin 0.05% / EDTA 0.02% in PBS, w/o: Ca and Mg, w: Phenol red is a porcine pancreas-derived enzyme plus EDTA in a PBS solution. Trypsin-EDTA solution is commonly used for the dissociation of anchorage-dependent (adherent) mammalian cells and tissues from culture surfaces. The concentration of trypsin necessary to dislodge cells from their substrate is dependent on the cell type and the age of the culture. Ethylenediaminetetraacetic acid (EDTA), a chelating agent, is often added to trypsin solutions to enhance enzymatic activity by neutralizing calcium and magnesium ions that enhance cell-to-cell adhesion and obscure the peptide bonds on which trypsin acts. LIQUID/POWDER: Liquid. PHENOL RED: With Phenol Red. Ca/Mg CONTENT: Without Ca and Mg. TRYPSIN: 0.05% Trypsin. EDTA: 0.02% EDTA. STERILE: Yes. STORAGE TEMP.: -20°C. MANUFACTURER: PAN-Biotech GmbH (Germany).
Catalog No. 50-273-3676