Learn More
Bellco Glass, Inc. Trypsinizing Flask,1 L
SDP
Catalog No. 501945235
Encompass_Preferred
Manufactured from type 1 class a Borosilicate glass. Designed with deep side baffles for optimal agitation. Large bottom surface for easy access with Bellco large orifice pipette. MADE IN THE USA.Break resistant lipSize: 1LOD x OAH – 128 x 223mm2 per case
Catalog No. 50-194-5235 Supplier Bellco Glass, Inc. Supplier No. 199201000
Manufactured from type 1 class a Borosilicate glass. Designed with deep side baffles for optimal agitation. Large bottom surface for easy access with Bellco large orifice pipette. MADE IN THE USA.Break resistant lipSize: 1LOD x OAH – 128 x 223mm2 per case