Learn More
Bellco Glass, Inc. Trypsinizing Flask,125mL
SDP
Catalog No. 501945228
Encompass_Preferred
Manufactured from Type 1 Borosilicate Glass. Designed with deep side baffles for optimal agitation. Promotes enzyme digestion of tissue fragments and homogenates into cell suspense. Includes rubber lined black phenolic screw caps. Integral dam contains non-trypsinized tissue during decanting. MADE IN THE USA.Specifications:Type 1 Class A Borosilicate GlassSize: 125mlSidearm Cap: 16mmAcid Etched & Filled GraduationsScrew Cap: 24mmOD X OAH (mm): 66 X 135Case Qty:3
Catalog No. 50-194-5228 Supplier Bellco Glass, Inc. Supplier No. 198900125
Manufactured from Type 1 Borosilicate Glass. Designed with deep side baffles for optimal agitation. Promotes enzyme digestion of tissue fragments and homogenates into cell suspense. Includes rubber lined black phenolic screw caps. Integral dam contains non-trypsinized tissue during decanting. MADE IN THE USA.Specifications:Type 1 Class A Borosilicate GlassSize: 125mlSidearm Cap: 16mmAcid Etched & Filled GraduationsScrew Cap: 24mmOD X OAH (mm): 66 X 135Case Qty:3