Learn More
Bellco Glass, Inc. Trypsinizing Flask,250mL
SDP
Supplier:  Bellco Glass, Inc. 199200250
Encompass_Preferred
Manufactured from type 1 class a Borosilicate glass. Designed with deep side baffles for optimal agitation. Large bottom surface for easy access with Bellco large orifice pipette. MADE IN THE USA.Break resistant lipSize: 250mLOD x OAH – 80 x 140mm3 per case
Catalog No. 50-194-5233